Product Details

SNP ID
rs12916714
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:77050818 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATTACTGCTATGCAGAGCTACAGA[C/G]TAAATGCTTAGAAGGGCAAAGACCA
Phenotype
MIM: 613134
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TSPAN3 PubMed Links

Gene Details

Gene
TSPAN3
Gene Name
tetraspanin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168412.1 Intron NP_001161884.1
NM_005724.5 Intron NP_005715.1
NM_198902.2 Intron NP_944492.1
XM_017021857.1 Intron XP_016877346.1

View Full Product Details