Product Details

SNP ID
rs11076155
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56451642 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGAAGCGGCCAGCGGAGCCCGG[C/G]CCAGCCCGGGTGGGAAAAAAGGGAA
Phenotype
MIM: 604978 MIM: 615857
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NUDT21 PubMed Links

Gene Details

Gene
NUDT21
Gene Name
nudix hydrolase 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007006.2 163 Intron NP_008937.1
Gene
OGFOD1
Gene Name
2-oxoglutarate and iron dependent oxygenase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324357.1 163 Silent Mutation GGC,GGG G10G NP_001311286.1
NM_001324358.1 163 Intron NP_001311287.1
NM_001324359.1 163 Intron NP_001311288.1
NM_001324360.1 163 UTR 5 NP_001311289.1
NM_001324361.1 163 Intron NP_001311290.1
NM_001324362.1 163 Intron NP_001311291.1
NM_001324363.1 163 Silent Mutation GGC,GGG G10G NP_001311292.1
NM_018233.3 163 Silent Mutation GGC,GGG G10G NP_060703.3

View Full Product Details