Product Details
- SNP ID
-
rs4073289
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:23184822 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGGGGCACATCGAAGTCACAGACA[C/T]GGGTCCTCTCTGGTCCTGGGGACCA
- Phenotype
-
MIM: 600761
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SCNN1G
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79474885] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SCNN1G
- Gene Name
- sodium channel epithelial 1 gamma subunit
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001039.3 |
|
Intron |
|
|
NP_001030.2 |
View Full Product Details