Product Details

SNP ID
rs10003054
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:158776594 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATAAAATATGATTTAAAGATAGT[A/T]ACAAAAATACCTTTTAAATTCAATG
Phenotype
MIM: 612768
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FNIP2 PubMed Links

Gene Details

Gene
FNIP2
Gene Name
folliculin interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323916.1 Intron NP_001310845.1
NM_020840.2 Intron NP_065891.1
XM_005263156.1 Intron XP_005263213.1
XM_005263158.1 Intron XP_005263215.1
XM_005263160.3 Intron XP_005263217.1
XM_011532154.2 Intron XP_011530456.1
XM_017008487.1 Intron XP_016863976.1

View Full Product Details