Product Details
- SNP ID
-
rs12679849
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:23136621 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTGTAATGGCGCTTGAAAGAGAAG[C/G]TCTTCGCAGTCTCGGATCTAACTTA
- Phenotype
-
MIM: 603614
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TNFRSF10D
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112453929,rs113448633] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TNFRSF10D
- Gene Name
- TNF receptor superfamily member 10d
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003840.4 |
2505 |
UTR 3 |
|
|
NP_003831.2 |
View Full Product Details