Product Details
- SNP ID
-
rs7027743
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:113153344 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTGACCGCCTAGATTTCCATTTTGA[A/G]GCTCTCATATAATTTACTTGACCAG
- Phenotype
-
MIM: 603088
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC31A2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112831964] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC31A2
- Gene Name
- solute carrier family 31 member 2
View Full Product Details