Product Details

SNP ID
rs11991303
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143791931 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGAGGTCTGGGGGGACAAGAAGC[A/G]GGCATTGGAAGCAGCCCATGCGGCA
Phenotype
MIM: 607733
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LOC642574 PubMed Links

Gene Details

Gene
LOC642574
Gene Name
IQ motif and ankyrin repeat domain-containing protein LOC642574
There are no transcripts associated with this gene.

Gene
SCRIB
Gene Name
scribbled planar cell polarity protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015356.4 Intron NP_056171.3
NM_182706.4 Intron NP_874365.3

View Full Product Details