Product Details
- SNP ID
-
rs2322143
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:71776722 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTATACACTCGGTAGATGCTTGTC[A/C]GTTTCTTGACTGGCCCACTCACTTC
- Phenotype
-
MIM: 607002
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PROK2
PubMed Links
Gene Details
- Gene
- PROK2
- Gene Name
- prokineticin 2
View Full Product Details