Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001080531.1 | 4871 | Intron | NP_001074000.1 | ||
XM_006714289.3 | 4871 | Intron | XP_006714352.1 | ||
XM_011532196.2 | 4871 | Intron | XP_011530498.1 | ||
XM_011532197.2 | 4871 | Intron | XP_011530499.1 | ||
XM_017008547.1 | 4871 | Intron | XP_016864036.1 | ||
XM_017008548.1 | 4871 | Intron | XP_016864037.1 | ||
XM_017008549.1 | 4871 | Intron | XP_016864038.1 | ||
XM_017008550.1 | 4871 | Intron | XP_016864039.1 | ||
XM_017008551.1 | 4871 | Intron | XP_016864040.1 | ||
XM_017008552.1 | 4871 | Intron | XP_016864041.1 | ||
XM_017008553.1 | 4871 | Intron | XP_016864042.1 | ||
XM_017008554.1 | 4871 | Intron | XP_016864043.1 | ||
XM_017008555.1 | 4871 | Intron | XP_016864044.1 | ||
XM_017008556.1 | 4871 | Intron | XP_016864045.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001306215.1 | 4871 | UTR 3 | NP_001293144.1 | ||
NM_178835.4 | 4871 | UTR 3 | NP_849157.2 | ||
XM_011531634.2 | 4871 | UTR 3 | XP_011529936.1 | ||
XM_011531635.2 | 4871 | UTR 3 | XP_011529937.1 | ||
XM_011531645.2 | 4871 | UTR 3 | XP_011529947.1 | ||
XM_017007768.1 | 4871 | UTR 3 | XP_016863257.1 | ||
XM_017007769.1 | 4871 | UTR 3 | XP_016863258.1 | ||
XM_017007770.1 | 4871 | UTR 3 | XP_016863259.1 | ||
XM_017007771.1 | 4871 | Intron | XP_016863260.1 | ||
XM_017007772.1 | 4871 | Intron | XP_016863261.1 | ||
XM_017007773.1 | 4871 | Intron | XP_016863262.1 | ||
XM_017007774.1 | 4871 | Intron | XP_016863263.1 | ||
XM_017007775.1 | 4871 | Intron | XP_016863264.1 | ||
XM_017007776.1 | 4871 | Intron | XP_016863265.1 | ||
XM_017007777.1 | 4871 | UTR 3 | XP_016863266.1 |