Product Details

SNP ID
rs1044527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:70367102 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAGCTCTCGCCCCTTTCATTCTCC[C/G]CCTGCAAGAACAAAGGCAAAATGAA
Phenotype
MIM: 609411
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
COX16 PubMed Links

Gene Details

Gene
COX16
Gene Name
COX16, cytochrome c oxidase assembly homolog
There are no transcripts associated with this gene.

Gene
SYNJ2BP
Gene Name
synaptojanin 2 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018373.2 6454 UTR 3 NP_060843.2
Gene
SYNJ2BP-COX16
Gene Name
SYNJ2BP-COX16 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202547.2 6454 Intron NP_001189476.1
NM_001202548.2 6454 Intron NP_001189477.1
NM_001202549.2 6454 Intron NP_001189478.1

View Full Product Details