Product Details

SNP ID
rs10251765
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:130083867 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTTTTCCCTTTTGGCTTCCTGGG[G/T]GTCAGGTCTCGTTTTCTTATGTGTC
Phenotype
MIM: 615152
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KLHDC10 PubMed Links

Gene Details

Gene
KLHDC10
Gene Name
kelch domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014997.3 Intron NP_055812.1
XM_005250233.4 Intron XP_005250290.1

View Full Product Details