Product Details
- SNP ID
-
rs7187575
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:28978780 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGTACCCGTCCTTAGCTTCAGGTG[C/T]CTTTGGTAGACACATAAACTCCCAT
- Phenotype
-
MIM: 602354
MIM: 612583
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LAT
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116364234] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LAT
- Gene Name
- linker for activation of T-cells
There are no transcripts associated with this gene.
- Gene
- SPNS1
- Gene Name
- sphingolipid transporter 1 (putative)
View Full Product Details