Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001286445.1 | 4520 | UTR 3 | NP_001273374.1 | ||
NM_001286446.1 | 4520 | Intron | NP_001273375.1 | ||
NM_001286447.1 | 4520 | Intron | NP_001273376.1 | ||
NM_014722.3 | 4520 | UTR 3 | NP_055537.2 | ||
NM_015864.3 | 4520 | Intron | NP_056948.2 | ||
XM_006715275.2 | 4520 | UTR 3 | XP_006715338.1 | ||
XM_006715277.1 | 4520 | UTR 3 | XP_006715340.1 | ||
XM_006715279.2 | 4520 | UTR 3 | XP_006715342.1 | ||
XM_006715281.3 | 4520 | Intron | XP_006715344.1 | ||
XM_011515007.1 | 4520 | UTR 3 | XP_011513309.1 | ||
XM_011515008.2 | 4520 | UTR 3 | XP_011513310.1 | ||
XM_011515009.1 | 4520 | UTR 3 | XP_011513311.1 | ||
XM_011515010.2 | 4520 | UTR 3 | XP_011513312.1 | ||
XM_011515012.1 | 4520 | UTR 3 | XP_011513314.1 | ||
XM_017011523.1 | 4520 | UTR 3 | XP_016867012.1 | ||
XM_017011524.1 | 4520 | Intron | XP_016867013.1 | ||
XM_017011525.1 | 4520 | Intron | XP_016867014.1 | ||
XM_017011526.1 | 4520 | Intron | XP_016867015.1 | ||
XM_017011527.1 | 4520 | Intron | XP_016867016.1 |