Product Details
- SNP ID
-
rs10791141
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:131375038 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCCACATTGCTCTCTCTCTTGCTG[C/T]AGCAGAGATCCTACAAATGCTTCAC
- Phenotype
-
MIM: 607938
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NTM
PubMed Links
Gene Details
- Gene
- NTM
- Gene Name
- neurotrimin
View Full Product Details