Product Details

SNP ID
rs7638808
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:52538040 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AAGAATGGATCCCAAGGATTGAGTC[A/G]TTTTAGGGAGAGGCTTTAACATGTG
Phenotype
MIM: 606083
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NT5DC2 PubMed Links

Gene Details

Gene
NT5DC2
Gene Name
5'-nucleotidase domain containing 2
There are no transcripts associated with this gene.

Gene
PBRM1
Gene Name
polybromo 1
There are no transcripts associated with this gene.

Gene
SMIM4
Gene Name
small integral membrane protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001124767.1 Intron NP_001118239.1
XM_011533729.2 Intron XP_011532031.1

View Full Product Details