Product Details

SNP ID
rs4628351
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:127433954 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGAGTCCTTAAAACCACCTCTCCAT[G/T]CCTCCATCTCAAGACACAGTTGCCT
Phenotype
MIM: 194552
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF79 PubMed Links

Gene Details

Gene
ZNF79
Gene Name
zinc finger protein 79
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286696.1 Intron NP_001273625.1
NM_001286697.1 Intron NP_001273626.1
NM_001286698.1 Intron NP_001273627.1
NM_001322260.1 Intron NP_001309189.1
NM_007135.2 Intron NP_009066.2
XM_006717279.3 Intron XP_006717342.1

View Full Product Details