Product Details

SNP ID
rs9902354
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75531479 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGGGCCGGCTGCTTTGAGCAGC[A/G]CTGGCAGCCTGGACTCATTTGTCCT
Phenotype
MIM: 608755
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LLGL2 PubMed Links

Gene Details

Gene
LLGL2
Gene Name
LLGL2, scribble cell polarity complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015002.1 Intron NP_001015002.1
NM_001031803.1 Intron NP_001026973.1
NM_004524.2 Intron NP_004515.2
XM_011524801.1 Intron XP_011523103.1
XM_011524802.1 Intron XP_011523104.1
XM_011524807.2 Intron XP_011523109.1
XM_017024625.1 Intron XP_016880114.1
XM_017024626.1 Intron XP_016880115.1
XM_017024627.1 Intron XP_016880116.1
XM_017024628.1 Intron XP_016880117.1
XM_017024629.1 Intron XP_016880118.1
XM_017024630.1 Intron XP_016880119.1
XM_017024631.1 Intron XP_016880120.1
XM_017024632.1 Intron XP_016880121.1
Gene
TSEN54
Gene Name
tRNA splicing endonuclease subunit 54
There are no transcripts associated with this gene.

View Full Product Details