Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001282949.2 | Intron | NP_001269878.1 | |||
NM_001282950.2 | Intron | NP_001269879.1 | |||
NM_001282951.2 | Intron | NP_001269880.1 | |||
NM_001282952.2 | Intron | NP_001269881.1 | |||
NM_001308381.1 | Intron | NP_001295310.1 | |||
NM_022730.3 | Intron | NP_073567.1 | |||
XM_006712693.1 | Intron | XP_006712756.1 | |||
XM_011511638.1 | Intron | XP_011509940.1 | |||
XM_011511639.1 | Intron | XP_011509941.1 | |||
XM_011511640.1 | Intron | XP_011509942.1 | |||
XM_017004716.1 | Intron | XP_016860205.1 | |||
XM_017004717.1 | Intron | XP_016860206.1 |