Product Details

SNP ID
rs10900482
Assay Type
Functionally tested
NCBI dbSNP Submissions
50
Location
Chr.1:205336974 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGTTTGGAACCCCCAATCATTCT[C/T]CATTTTTCATCCTAAGCTACCTTAA
Phenotype
MIM: 614503
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHDC8A PubMed Links

Gene Details

Gene
KLHDC8A
Gene Name
kelch domain containing 8A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271863.1 2022 UTR 3 NP_001258792.1
NM_001271864.1 2022 UTR 3 NP_001258793.1
NM_001271865.1 2022 UTR 3 NP_001258794.1
NM_018203.2 2022 UTR 3 NP_060673.1

View Full Product Details