Product Details

SNP ID
rs11598629
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:5685287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTGGCTGCGAGGGTCGACGCGTT[C/T]TCCTTGCCTTTTGCCGCGCTCCGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM208B PubMed Links

Gene Details

Gene
FAM208B
Gene Name
family with sequence similarity 208 member B
There are no transcripts associated with this gene.

View Full Product Details