Product Details

SNP ID
rs12603583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:80270438 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAATGCAAATCTCTTCAAATCCCCT[C/G]CACAAAGCTTCTCTGGCCACTCACT
Phenotype
MIM: 613768
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RNF213 PubMed Links

Gene Details

Gene
RNF213
Gene Name
ring finger protein 213
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256071.2 Intron NP_001243000.2
NM_020954.3 Intron NP_066005.2
XM_005257545.4 Intron XP_005257602.2
XM_005257546.4 Intron XP_005257603.2
XM_006721995.3 Intron XP_006722058.1
XM_011525084.2 Intron XP_011523386.1
XM_011525086.2 Intron XP_011523388.1
XM_011525087.2 Intron XP_011523389.1
XM_017024905.1 Intron XP_016880394.1

View Full Product Details