Product Details

SNP ID
rs191294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:134525235 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCGCGTAGGGGGCGGGGTGTGAGCA[A/G]AAAGTGTGAGTGAAAGAGTGGAGGG
Phenotype
MIM: 610515
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
JADE2 PubMed Links

Gene Details

Gene
JADE2
Gene Name
jade family PHD finger 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289984.1 Intron NP_001276913.1
NM_001289985.1 Intron NP_001276914.1
NM_001308143.1 Intron NP_001295072.1
NM_015288.5 Intron NP_056103.4
XM_005271945.1 Intron XP_005272002.1
XM_005271946.1 Intron XP_005272003.1
XM_005271948.4 Intron XP_005272005.1
XM_011543291.2 Intron XP_011541593.1
XM_017009282.1 Intron XP_016864771.1

View Full Product Details