Product Details

SNP ID
rs152617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:14587931 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCAGTCTCCTTGATATTCCCTT[G/T]TCCTCCCCATTTAGCAGCCTTTTTG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM105A PubMed Links

Gene Details

Gene
FAM105A
Gene Name
family with sequence similarity 105 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019018.2 Intron NP_061891.1
XM_011514056.2 Intron XP_011512358.1

View Full Product Details