Product Details

SNP ID
rs351780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:28346420 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGACAGGCACGGGATGGCAGCAC[G/T]GGTGGAAGGGAGGCAAGGAGGCCGC
Phenotype
MIM: 601459 MIM: 609494
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PNOC PubMed Links

Gene Details

Gene
PNOC
Gene Name
prepronociceptin
There are no transcripts associated with this gene.

Gene
ZNF395
Gene Name
zinc finger protein 395
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018660.2 3967 UTR 3 NP_061130.1

View Full Product Details