Product Details

SNP ID
rs602946
Assay Type
DME
NCBI dbSNP Submissions
11
Location
Chr.1:20589042 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
CTGGCTGCAGGGACACACCCAAGGG[G/A]AGGAGCTGCAATCGTGTCTGGGGCC
Phenotype
MIM: 123920
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CDA PubMed Links

Gene Details

Gene
CDA
Gene Name
cytidine deaminase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001785.2 92 UTR 5 NP_001776.1

View Full Product Details