Product Details

SNP ID
rs263437
Assay Type
Validated
NCBI dbSNP Submissions
30
Location
Chr.1:86705274 on Build GRCh38
Set Membership
JSNP Validated
Context Sequence [VIC/FAM]
CGCCTTTCTGATACCACGACGTCAG[C/T]GCTCCTGTGGCCTCCACCCCGGGGC
Phenotype
MIM: 609287
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105378828 PubMed Links

Gene Details

Gene
LOC105378828
Gene Name
uncharacterized LOC105378828
There are no transcripts associated with this gene.

Gene
SH3GLB1
Gene Name
SH3 domain containing GRB2 like endophilin B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206651.1 Intron NP_001193580.1
NM_001206652.1 Intron NP_001193581.1
NM_001206653.1 Intron NP_001193582.1
NM_016009.4 Intron NP_057093.1
XM_006710672.2 Intron XP_006710735.1
XM_006710673.2 Intron XP_006710736.1
XM_011541538.1 Intron XP_011539840.1
XM_017001408.1 Intron XP_016856897.1
XM_017001409.1 Intron XP_016856898.1
XM_017001410.1 Intron XP_016856899.1

View Full Product Details