Product Details
- SNP ID
-
rs376165
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:139535574 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCCTTTCTCTTCCTTTTTGGAGTTA[A/G]TCAGGAAGTAGTCCCAAATACCCCA
- Phenotype
-
MIM: 300746
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
F9
PubMed Links
Gene Details
- Gene
- F9
- Gene Name
- coagulation factor IX
View Full Product Details