Product Details

SNP ID
rs251913
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39307076 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCAAATAGGGAAAGACAGGCACTA[C/T]AGAGAGACGACAAGAGGGCGGGATA
Phenotype
MIM: 607403 MIM: 612807
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFNL1 PubMed Links

Gene Details

Gene
IFNL1
Gene Name
interferon lambda 1
There are no transcripts associated with this gene.

Gene
LRFN1
Gene Name
leucine rich repeat and fibronectin type III domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020862.1 2873 UTR 3 NP_065913.1
XM_005259102.4 2873 UTR 3 XP_005259159.1
XM_017027033.1 2873 UTR 3 XP_016882522.1

View Full Product Details