Product Details

SNP ID
rs40501
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11425820 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTGAGGGGGCAGGGCCTTAGGCAA[C/T]GGTTGGCTCAGAGGCACGGTCAGAT
Phenotype
MIM: 615956 MIM: 177060 MIM: 616743
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC151 PubMed Links

Gene Details

Gene
CCDC151
Gene Name
coiled-coil domain containing 151
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302453.1 Intron NP_001289382.1
NM_001302454.1 Intron NP_001289383.1
NM_145045.4 Intron NP_659482.3
XM_017026241.1 Intron XP_016881730.1
Gene
PRKCSH
Gene Name
protein kinase C substrate 80K-H
There are no transcripts associated with this gene.

Gene
RGL3
Gene Name
ral guanine nucleotide dissociation stimulator like 3
There are no transcripts associated with this gene.

View Full Product Details