Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001302453.1 | 39 | Missense Mutation | AAA,GAA | K10E | NP_001289382.1 |
NM_001302454.1 | 39 | Intron | NP_001289383.1 | ||
NM_145045.4 | 39 | Intron | NP_659482.3 | ||
XM_017026241.1 | 39 | Intron | XP_016881730.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001001329.2 | 39 | Intron | NP_001001329.1 | ||
NM_001289102.1 | 39 | UTR 5 | NP_001276031.1 | ||
NM_001289103.1 | 39 | Intron | NP_001276032.1 | ||
NM_001289104.1 | 39 | Intron | NP_001276033.1 | ||
NM_002743.3 | 39 | Intron | NP_002734.2 | ||
XM_011528130.1 | 39 | Intron | XP_011526432.1 | ||
XM_011528131.1 | 39 | Intron | XP_011526433.1 | ||
XM_011528132.1 | 39 | Intron | XP_011526434.1 | ||
XM_017026977.1 | 39 | Intron | XP_016882466.1 |