Product Details

SNP ID
rs223427
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:102800912 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGTTACATATAACGTAACAGAT[A/G]CCTATTAAGAGACCAGAATTGATCT
Phenotype
MIM: 602963
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC102723704 PubMed Links

Gene Details

Gene
LOC102723704
Gene Name
uncharacterized LOC102723704
There are no transcripts associated with this gene.

Gene
UBE2D3
Gene Name
ubiquitin conjugating enzyme E2 D3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300795.1 Intron NP_001287724.1
NM_003340.6 Intron NP_003331.1
NM_181886.3 Intron NP_871615.1
NM_181887.2 Intron NP_871616.1
NM_181888.3 Intron NP_871617.1
NM_181889.2 Intron NP_871618.1
NM_181890.2 Intron NP_871619.1
NM_181891.2 Intron NP_871620.1
NM_181892.3 Intron NP_871621.1
NM_181893.2 Intron NP_871622.1
XM_017008584.1 Intron XP_016864073.1

View Full Product Details