Product Details
- SNP ID
-
rs514949
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:113907892 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGGGTCCAGACATGTAAGTGAAGAA[A/G]CCGTGTTGGATGTTGAGCTTGGTTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ34503
PubMed Links
Gene Details
- Gene
- FLJ34503
- Gene Name
- uncharacterized FLJ34503
There are no transcripts associated with this gene.
View Full Product Details