Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000244.3 | Intron | NP_000235.2 | |||
NM_130799.2 | Intron | NP_570711.1 | |||
NM_130800.2 | Intron | NP_570712.1 | |||
NM_130801.2 | Intron | NP_570713.1 | |||
NM_130802.2 | Intron | NP_570714.1 | |||
NM_130803.2 | Intron | NP_570715.1 | |||
NM_130804.2 | Intron | NP_570716.1 | |||
XM_005274001.4 | Intron | XP_005274058.1 | |||
XM_011545040.1 | Intron | XP_011543342.1 | |||
XM_011545041.2 | Intron | XP_011543343.1 | |||
XM_011545042.2 | Intron | XP_011543344.1 | |||
XM_017017765.1 | Intron | XP_016873254.1 | |||
XM_017017766.1 | Intron | XP_016873255.1 | |||
XM_017017767.1 | Intron | XP_016873256.1 | |||
XM_017017768.1 | Intron | XP_016873257.1 | |||
XM_017017769.1 | Intron | XP_016873258.1 | |||
XM_017017770.1 | Intron | XP_016873259.1 |