Product Details
- SNP ID
-
rs578506
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:121452768 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTTTTTTCCTTCACGAGTACAACC[C/G]TCAGCACTTGAATCGCATTGATCTT
- Phenotype
-
MIM: 602005
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC105369536
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111342908] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC105369536
- Gene Name
- uncharacterized LOC105369536
There are no transcripts associated with this gene.
- Gene
- SORL1
- Gene Name
- sortilin related receptor 1
View Full Product Details