Product Details

SNP ID
rs380691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:80656215 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTTGAGTTGCTGAGGCTCCTGACT[A/G]TCAGACTCTACCTTGTCTTTTATGG
Phenotype
MIM: 126060 MIM: 600887
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHFR PubMed Links

Gene Details

Gene
DHFR
Gene Name
dihydrofolate reductase
There are no transcripts associated with this gene.

Gene
MSH3
Gene Name
mutS homolog 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002439.4 Intron NP_002430.3
Gene
MTRNR2L2
Gene Name
MT-RNR2-like 2
There are no transcripts associated with this gene.

View Full Product Details