Product Details

SNP ID
rs299867
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:54551398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATGAAGTGGTCAGCCTGCTGCCA[C/T]GCTCGGGGCTGGGAGAACTGCTGAG
Phenotype
MIM: 606803
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACOT11 PubMed Links

Gene Details

Gene
ACOT11
Gene Name
acyl-CoA thioesterase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015547.3 Intron NP_056362.1
NM_147161.3 Intron NP_671517.1

View Full Product Details