Product Details

SNP ID
rs480210
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:155217737 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCACTTTCTTAATGATTTTACAAAA[C/T]TCGAGGTGGTAGCTACTACTGTTAT
Phenotype
MIM: 300133
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
VBP1 PubMed Links

Gene Details

Gene
VBP1
Gene Name
VHL binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303543.1 Intron NP_001290472.1
NM_001303544.1 Intron NP_001290473.1
NM_001303545.1 Intron NP_001290474.1
NM_003372.6 Intron NP_003363.1

View Full Product Details