Product Details

SNP ID
rs184164
Assay Type
Functionally tested
NCBI dbSNP Submissions
33
Location
Chr.1:200895139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGGGGCTGAGAGCAGGCTGGCC[C/T]GGGAGTGGAGGAGCCCAAGCAGTGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf106 PubMed Links

Gene Details

Gene
C1orf106
Gene Name
chromosome 1 open reading frame 106
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142569.2 Intron NP_001136041.1
NM_018265.3 Intron NP_060735.3
XM_011509754.2 Intron XP_011508056.1
XM_011509755.1 Intron XP_011508057.1
XM_011509756.2 Intron XP_011508058.1

View Full Product Details