Product Details

SNP ID
rs677088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:2937558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTCACAGTTTTACATGGCAAACA[A/G]AAATTTCTGCAAACCCCCTGCTATT
Phenotype
MIM: 608397
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSMD1 PubMed Links

Gene Details

Gene
CSMD1
Gene Name
CUB and Sushi multiple domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033225.5 9219 UTR 3 NP_150094.5
XM_011534752.2 9219 UTR 3 XP_011533054.1
XM_011534753.2 9219 UTR 3 XP_011533055.1
XM_011534754.1 9219 UTR 3 XP_011533056.1
XM_017013731.1 9219 Intron XP_016869220.1

View Full Product Details