Product Details
- SNP ID
-
rs546502
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:70272383 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTAACCCAGGGGAAGTTACACGAG[A/G]TTGGTCATGAAAGAGATTCCTTGCA
- Phenotype
-
MIM: 611054
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PPFIA1
PubMed Links
Gene Details
- Gene
- PPFIA1
- Gene Name
- PTPRF interacting protein alpha 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003626.3 |
439 |
Missense Mutation |
ATT,GTT |
I71V |
NP_003617.1 |
NM_177423.2 |
439 |
Missense Mutation |
ATT,GTT |
I71V |
NP_803172.1 |
XM_006718716.3 |
439 |
Missense Mutation |
ATT,GTT |
I71V |
XP_006718779.1 |
XM_011545306.2 |
439 |
Missense Mutation |
ATT,GTT |
I71V |
XP_011543608.1 |
XM_011545307.2 |
439 |
Missense Mutation |
ATT,GTT |
I71V |
XP_011543609.1 |
XM_011545308.2 |
439 |
Missense Mutation |
ATT,GTT |
I71V |
XP_011543610.1 |
XM_011545309.2 |
439 |
Missense Mutation |
ATT,GTT |
I71V |
XP_011543611.1 |
XM_011545310.2 |
439 |
Missense Mutation |
ATT,GTT |
I71V |
XP_011543612.1 |
XM_011545311.2 |
439 |
Missense Mutation |
ATT,GTT |
I71V |
XP_011543613.1 |
XM_011545312.2 |
439 |
Missense Mutation |
ATT,GTT |
I71V |
XP_011543614.1 |
XM_011545313.2 |
439 |
Missense Mutation |
ATT,GTT |
I71V |
XP_011543615.1 |
XM_011545314.2 |
439 |
Missense Mutation |
ATT,GTT |
I71V |
XP_011543616.1 |
XM_011545315.2 |
439 |
Missense Mutation |
ATT,GTT |
I71V |
XP_011543617.1 |
XM_011545316.2 |
439 |
Missense Mutation |
ATT,GTT |
I71V |
XP_011543618.1 |
XM_011545317.2 |
439 |
Missense Mutation |
ATT,GTT |
I71V |
XP_011543619.1 |
XM_011545318.2 |
439 |
Intron |
|
|
XP_011543620.1 |
XM_011545319.1 |
439 |
Missense Mutation |
ATT,GTT |
I71V |
XP_011543621.1 |
XM_017018448.1 |
439 |
Missense Mutation |
ATT,GTT |
I71V |
XP_016873937.1 |
XM_017018449.1 |
439 |
Missense Mutation |
ATT,GTT |
I71V |
XP_016873938.1 |
XM_017018450.1 |
439 |
Missense Mutation |
ATT,GTT |
I71V |
XP_016873939.1 |
View Full Product Details