Product Details
- SNP ID
-
rs513342
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:69348408 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTAGCACTCTGACCTAGCAGTCAA[C/T]ATGAAGGCTCTCATTGTTCTGGGGC
- Phenotype
-
MIM: 153450
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LYZ
PubMed Links
Gene Details
- Gene
- LYZ
- Gene Name
- lysozyme
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000239.2 |
55 |
UTR 5 |
|
|
NP_000230.1 |
View Full Product Details