Product Details

SNP ID
rs121913547
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:69350192 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGACAGAAGCACTGATTATGGGA[C/T]ATTTCAGATCAATAGCCGCTACTGG
Phenotype
MIM: 153450 MIM: 602116
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYZ PubMed Links

Gene Details

Gene
LYZ
Gene Name
lysozyme
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000239.2 276 Missense Mutation ACA,ATA T74I NP_000230.1
Gene
YEATS4
Gene Name
YEATS domain containing 4
There are no transcripts associated with this gene.

View Full Product Details