Product Details

SNP ID
rs680199
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:36684660 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATCATTCTTCTGCCATGATGTGTC[C/T]CTGCATCCCAGCTCAAACCATTGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERTM1 PubMed Links

Gene Details

Gene
SERTM1
Gene Name
serine rich and transmembrane domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203451.2 Intron NP_982276.2

View Full Product Details