Product Details
- SNP ID
-
rs682626
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:6774058 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AGATGGGCCCATGGGGGTTCCTGGC[A/G]GGTGGTTTTCAGGGGCAGGGCCATT
- Phenotype
-
MIM: 604721
MIM: 164875
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SH2D3A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs146242402] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SH2D3A
- Gene Name
- SH2 domain containing 3A
There are no transcripts associated with this gene.
- Gene
- VAV1
- Gene Name
- vav guanine nucleotide exchange factor 1
View Full Product Details