Product Details

SNP ID
rs678746
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:223877276 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGAATGACCCTATTGTCCTGGTA[C/T]GATCCCAACACAGTGATGCAGTACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WDFY1 PubMed Links

Gene Details

Gene
WDFY1
Gene Name
WD repeat and FYVE domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020830.4 2731 UTR 3 NP_065881.1

View Full Product Details