Product Details
- SNP ID
-
rs584358
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:223878021 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- TTTCACAACTTACTTAATGACAACC[C/T]CGAAAGTTCACCTGTGGCATACTGG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
WDFY1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114444291] are located under a probe and SNP(s) [rs115114882] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- WDFY1
- Gene Name
- WD repeat and FYVE domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020830.4 |
1986 |
UTR 3 |
|
|
NP_065881.1 |
View Full Product Details