Product Details

SNP ID
rs375117
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:2061775 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCTGAGCCACACCGGTTCCCCCA[C/T]GCCGGATGCCGCAGGGGTGAAGACT
Phenotype
MIM: 610647
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NAT8L PubMed Links

Gene Details

Gene
NAT8L
Gene Name
N-acetyltransferase 8 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178557.3 Intron NP_848652.2

View Full Product Details