Product Details
- SNP ID
-
rs272496
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
60
- Location
-
Chr.1:86352889 on Build GRCh38
- Set Membership
-
JSNP
- Context Sequence [VIC/FAM]
- TCATTTTGCAAACAAGTTGATTTTG[C/T]TCTTCATTTTTCTTTCTAAGCTCAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR7856
PubMed Links
Gene Details
- Gene
- MIR7856
- Gene Name
- microRNA 7856
There are no transcripts associated with this gene.
- Gene
- ODF2L
- Gene Name
- outer dense fiber of sperm tails 2 like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001007022.2 |
2184 |
Silent Mutation |
GAA,GAG |
E621E |
NP_001007023.2 |
NM_001184765.1 |
2184 |
Silent Mutation |
GAA,GAG |
E568E |
NP_001171694.1 |
NM_001184766.1 |
2184 |
Silent Mutation |
GAA,GAG |
E539E |
NP_001171695.1 |
NM_020729.2 |
2184 |
Silent Mutation |
GAA,GAG |
E605E |
NP_065780.2 |
XM_005271054.3 |
2184 |
Silent Mutation |
GAA,GAG |
E592E |
XP_005271111.1 |
XM_011541815.2 |
2184 |
Silent Mutation |
GAA,GAG |
E621E |
XP_011540117.1 |
XM_011541819.2 |
2184 |
Silent Mutation |
GAA,GAG |
E621E |
XP_011540121.1 |
XM_011541821.2 |
2184 |
Silent Mutation |
GAA,GAG |
E568E |
XP_011540123.1 |
XM_011541823.2 |
2184 |
Intron |
|
|
XP_011540125.1 |
XM_011541824.2 |
2184 |
Intron |
|
|
XP_011540126.1 |
XM_017001876.1 |
2184 |
Silent Mutation |
GAA,GAG |
E621E |
XP_016857365.1 |
XM_017001877.1 |
2184 |
Silent Mutation |
GAA,GAG |
E592E |
XP_016857366.1 |
XM_017001878.1 |
2184 |
Silent Mutation |
GAA,GAG |
E582E |
XP_016857367.1 |
XM_017001879.1 |
2184 |
Silent Mutation |
GAA,GAG |
E592E |
XP_016857368.1 |
XM_017001880.1 |
2184 |
Silent Mutation |
GAA,GAG |
E553E |
XP_016857369.1 |
XM_017001881.1 |
2184 |
Silent Mutation |
GAA,GAG |
E422E |
XP_016857370.1 |
XM_017001882.1 |
2184 |
Silent Mutation |
GAA,GAG |
E317E |
XP_016857371.1 |
View Full Product Details