Product Details
- SNP ID
-
rs558465
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:55572272 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTGCTAATCATTATTAGTGTCAAG[A/G]CCAGCCAGGCACTTAAGAACCCAAT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR4C16
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75881957] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR4C16
- Gene Name
- olfactory receptor family 4 subfamily C member 16 (gene/pseudogene)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004701.2 |
145 |
Missense Mutation |
ACC,GCC |
T49A |
NP_001004701.2 |
View Full Product Details