Product Details

SNP ID
rs225360
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.21:42313821 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TTTAAAGAGAGGCAGTCTGTTAAAT[A/G]CTCAGCTCGGGGACTCTGGCCATTT
Phenotype
MIM: 603076 MIM: 600633
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCG1 PubMed Links

Gene Details

Gene
ABCG1
Gene Name
ATP binding cassette subfamily G member 1
There are no transcripts associated with this gene.

Gene
TFF3
Gene Name
trefoil factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003226.3 Intron NP_003217.3

View Full Product Details